Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium meta-Silicate Nonahydrate (Crystalline/Certified), Fisher Chemical

Catalog No. S408500 Shop All Fisherbrand Products
Change view
Click to view available options
Poly Bottle; 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S408-500 Poly Bottle; 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S408-500

missing translation for 'validMainframeMsg'

Specifications

Melting Point 72°C
Color White
CAS 13517-24-3
pH 12.4
Physical Form Solid
Quantity 500 g
Molecular Formula H18Na2O12Si
MDL Number MFCD00149175
Synonym sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9
InChI Key PHIQPXBZDGYJOG-UHFFFAOYSA-N
SMILES O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]
IUPAC Name disodium;dioxido(oxo)silane;nonahydrate
Molecular Weight (g/mol) 284.197
PubChem CID 61639
Formula Weight 284.2
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Sodium meta-Silicate Nonahydrate
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.