Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium meta-Silicate Pentahydrate (Crystalline/Technical), Fisher Chemical

Catalog No. S4073 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
3 kg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S407-3 3 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S407-3

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10213-79-3
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.137
MDL Number MFCD00149176
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
Synonym sodium metasilicate pentahydrate, silicic acid, disodium salt, pentahydrate, disodium oxosilanediolate pentahydrate, silicic acid h2sio3 , disodium salt, pentahydrate, 2na.so3i.5h2o, sodium meta-silicate pentahydrate, sodium silicate solution water glass, disodium dioxido oxo silane pentahydrate, sodium metasilicate pentahydrate, tech., disodium oxosilanebis olate pentahydrate
PubChem CID 61495
IUPAC Name disodium;dioxido(oxo)silane;pentahydrate
SMILES O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Melting Point 72°C
Color White
pH 12.6
Physical Form Solid
Quantity 3 kg
Formula Weight 212.15
Grade Technical
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Sodium meta-Silicate Pentahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.