Learn More
Medchemexpress LLC Sodium metabisulfite | 7681-57-4 | MFCD00167602 | 1mL
SDP
Catalog No. 5000266601
Encompass_Preferred
Sodium metabisulfite a sulfite salt is used as an antioxidant and antimicrobial agent in a variety of drugs and functions as a preservative in many food preparations[1]
Catalog No. 50-002-66601 Supplier Medchemexpress LLC Supplier No. HYY132610MM/1ML
Sodium metabisulfite a sulfite salt is used as an antioxidant and antimicrobial agent in a variety of drugs and functions as a preservative in many food preparations[1]