Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Metabisulfite, MP Biomedicals™

Catalog No. MP021914381 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021914381 100 g
MP021914385 500 g
MP219143805 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP021914381 Supplier MP Biomedicals, Inc Supplier No. 02191438.1
Add to Cart
Add to Cart

Relied on by scientists. Sodium Metabisulfite, MP Biomedicals is ACS reagent grade with a purity greater than 97%. MP Biomedicals has delivered quality biochemicals for over 50 years.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 100 g
Formula Weight 190.11g/mol
Percent Purity ≥97.0%
Grade ACS Reagent
Recommended Storage Store at Room Temperature (15–30°C)
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.