Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium metabisulfite puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5% | Purity: 65.0-67.4% SO2 basis; 97-100.5% | Mol Wt: 190.11 | 7681-57-4 | MFCD00167602 | 1KG
Catalog No. 501764562
Encompass
Sodium metabisulfite puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5% | Purity: 65.0-67.4% SO2 basis; 97-100.5% | Mol Wt: 190.11 | 7681-57-4 | MFCD00167602 | 1KG
Catalog No. 50-176-4562 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 134591KGR
Sodium metabisulfite puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5% | Purity: 65.0-67.4% SO2 basis; 97-100.5% | Mol Wt: 190.11 | 7681-57-4 | MFCD00167602 | 1KG