Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium metaperiodate, 98%

Catalog No. p-4407911
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1379809 10 g
AA1379822 100 g
AA1379836 500 g
AA13798A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1379809 Supplier Thermo Scientific Chemicals Supplier No. 01379809
Only null left
Add to Cart

Source of Periodic Acid, As An Analytical Reagent, and As An Oxidizing Agent | CAS: 7790-28-5 | INaO4 | 213.89 g/mol

Source of periodic acid, as an analytical reagent, and as an oxidizing agent Sodium metaperiodate is used as an oxidizing reagent involved in the oxidation of cellulose. It is involved in the cleavage of vicinal diols to prepare two aldehydes. This oxidation is commonly used in selectively label RNA, saccharides and other compounds with vicinal diols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
MDL Number MFCD00003534
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym sodium periodate, sodium metaperiodate, sodium m-periodate, sodiumperiodate, sodium meta periodate, periodic acid, sodium salt, sodium meta-periodate, periodic acid hio4 , sodium salt, periodate sodium, sodium penodate
PubChem CID 23667635
ChEBI CHEBI:75226
IUPAC Name sodium;periodate
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Melting Point 300°C (decomposition)
Color Colorless
Physical Form Powder
Quantity 10 g
Assay Percent Range 98%
Solubility Information Soluble in water,sulfuric acid,nitric acid and acetic acid.
Formula Weight 213.89
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Appearance Colorless powder
Density 4.174 g/cm3
Chemical Name or Material Sodium metaperiodate
Show More Show Less
Hazard Category H271-H314-H335-H372
Precautionary Statement P210-P220-P221-P260-P264b-P270-P271-P280-P283-P301+P330+P331-P304+P340-P305+P351+P338-P306+P360-P310-P353-P363-P370+P378q-P371+P380+P375-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: I
EINECSNumber 232-197-6
RTECSNumber SD4550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.