Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium metaperiodate, 99.0%, for analysis EMSURE™ ACS, Reag. Ph Eur, MilliporeSigma™

Catalog No. M1065970050 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1065970050 50 g
M1065970250 250 g
M1065971000 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1065970050 Supplier MilliporeSigma Supplier No. 1.06597.0050
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym Sodium periodate
IUPAC Name sodium periodate
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Melting Point 270°C (decomposes)
pH 5.2 (50 g/L, H2O, 20°C)
Quantity 50 g
Assay Percent Range Assay (iodometric): ≥ 99.0%; Assay (out of dried substance): 99.8 to 100.3%
Solubility Information 91 g/L
Packaging Plastic Bottle
Density 3.87 g/cm3 (at 20°C); Bulk Density: 2900 kg/m3
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.