Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium metaperiodate, 99.0%, for analysis EMSURE™ ACS, Reag. Ph Eur, MilliporeSigma™

Catalog No. M1065971000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1065971000 1 kg
M1065970050 50 g
M1065970250 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1065971000 Supplier MilliporeSigma Supplier No. 1065971000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7790-28-5 | Formula: INaO4 | Weight: 213.89 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-28-5
Molecular Formula INaO4
Molecular Weight (g/mol) 213.89
InChI Key JQWHASGSAFIOCM-UHFFFAOYSA-M
Synonym Sodium periodate
IUPAC Name sodium periodate
SMILES [Na+].[O-][I](=O)(=O)=O

Specifications

Melting Point 270°C (decomposes)
pH 5.2 (50 g/L, H2O, 20°C)
Physical Form Crystals
Quantity 1 kg
Packaging Plastic Bottle
Assay Percent Range Assay (iodometric): ≥ 99.0%; Assay (out of dried substance): 99.8 to 100.3%
Density 3.87 g/cm3 (at 20°C); Bulk Density: 2900 kg/m3
Solubility Information 91 g/L
Percent Purity ≥99.0%
Grade ACS
Chemical Name or Material Sodium Metaperiodate
Show More Show Less
Recommended Storage Store at 5 to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.