Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Metasilicate

Catalog No. 30781525KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-781-525KG 2.5 kg
30-781-525G 25 g
30-781-51KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 30-781-525KG Supplier Sigma Aldrich Supplier No. 3078152.5KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium metasilicate (SMS) has been used in the preparation of polypyrrole/sodium metasilicate (PPy/Na2SiO3) and polythiophene/sodium metasilicate (PT/Na2SiO3) composites, novel high dielectric constant materials for use in power electronic systems.SMS can also be used:To prepare NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption and catalysis.As a low-cost alternative to tetraethyl orthosilicate (TEOS) to synthesize highly ordered mesoporous silicas.To prepare geopolymer concrete.

Specifications

CAS 6834-92-0
Quantity 2.5 kg
Molecular Formula Na2O3Si
Linear Formula Na2SiO3
MDL Number MFCD00003492
Synonym Disodium metasilicate; Disodium silicate; Sodium silicate; Sodium silicon oxide
Molecular Weight (g/mol) 122.06 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.