Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Metasilicate, MP Biomedicals

Catalog No. ICN19138291 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19138291 1 kg
ICN19138280 100 g
ICN19138283 250 g
ICN19138290 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19138291 Supplier MP Biomedicals, Inc Supplier No. 0219138291
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13517-24-3
Molecular Formula H18Na2O12Si
Molecular Weight (g/mol) 284.197
InChI Key PHIQPXBZDGYJOG-UHFFFAOYSA-N
Synonym sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9
PubChem CID 61639
IUPAC Name disodium;dioxido(oxo)silane;nonahydrate
SMILES O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Quantity 1 kg
Assay Percent Range ≥97%
Formula Weight 284.2
Grade Reagent
Chemical Name or Material Sodium Metasilicate
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.