Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium metasilicate nonahydrate

Catalog No. 503126002 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-6002 1 kg
50-312-6088 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-312-6002 Supplier Sigma Aldrich Supplier No. S43921KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Na2SiO3.9H2O can be used to prepare honeycomb porous silicon (hp-Si). Sodium metasilicate nonahydrate combined with cobalt chloride (CoCl2. 6H2O) may be used to prepare Co3O4 nanoparticles. The product may be used to impart flame retardant and antimicrobial properties in jute textiles.

Specifications

CAS 13517-24-3
Quantity 1 kg
Molecular Formula Na2O3Si -+ 9H2O
Linear Formula Na2O3Si -+ 9H2O
MDL Number MFCD00149175
Molecular Weight (g/mol) 284.2
Percent Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.