Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Metasilicate Pentahydrate

Catalog No. 717461KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-746-1KG 1 kg
71-746-250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 71-746-1KG Supplier Sigma Aldrich Supplier No. 717461KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium metasilicate pentahydrate can be used:As a silicate electrolyte in plasma electrolytic oxidation (PEO) coating.For the synthesis of sintered mullite ceramics (3Al2O3-+2SiO2) by the co-precipitation method.As a source of silica for the synthesis of silica-zirconia composites for the chromatographic applications.For the synthesis of photo-Fenton catalyst, nontronite, for the degradation of Rhodamine B.As a starting material for the synthesis of zeolite NaA layers on +a-Al2O3 supports.

Specifications

CAS 10213-79-3
Quantity 1 kg
Molecular Formula Na2O3Si -+ 5H2O
Linear Formula Na2SiO3 -+ 5H2O
MDL Number MFCD00149176
Molecular Weight (g/mol) 212.14 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.