Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Metasilicate Pentahydrate, Purified, Reagents

Catalog No. 01336976
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-976 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-976 Supplier Reagents Holdings Llc Supplier No. C2324800500C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Metasilicate Pentahydrate, Purified, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10213-79-3
Molecular Formula H10Na2O8Si
Molecular Weight (g/mol) 212.14
InChI Key ADPGKKZKGXANON-UHFFFAOYSA-N
IUPAC Name disodium oxosilanebis(olate) pentahydrate
SMILES O.O.O.O.O.[Na+].[Na+].[O-][Si]([O-])=O

Specifications

Boiling Point >1200°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.52
Chemical Name or Material Sodium Metasilicate Pentahydrate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.