Learn More
Selleck Chemical LLC SodiuM Metatungstate
SDP
Supplier:  Selleck Chemical LLC S552525MG
Encompass_Preferred
Sodium metatungstate (SMT) solution is an inorganic heavy liquid which is widely used in density fractionation Sodium metatungstate (Sodium polyoxotungstates) is a potent and selective inhibitor of ecto-nucleoside triphosphate diphosphohydrolase (NTPDases CD39) with Ki of 0 140 M 0 910 M and 0 563 M for NTPDase1 NTPDase2 and NTPDase3 respectively
Catalog No. 50-000-03167