Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium metatungstate | 12141-67-2 | MFCD00061466 | 93.4% | 2,968.0 g/mol | Na6O39W12 | 100 MG
SDP

Catalog No. 5000246228
Encompass_Preferred

Sodium metatungstate is an inorganic polytungstate reagent used in analytical chemistry, density separation, and various research applications. Supplied as a solid laboratory reagent, it is commonly used to prepare density-gradient media, to separate or concentrate biological specimens, and in some biochemical assays.

  • High-density, water-soluble polytungstate reagent.
  • Used for density gradient separation and sample separation.
  • Soluble in water and DMSO with sonication (reported solubility ~50 mg/mL).
  • Available in small laboratory pack sizes such as 100 MG.
  • CAS number 12141-67-2; reported purity varies by supplier.

Catalog No. 50-002-46228 Supplier Medchemexpress LLC Supplier No. HY103259100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium metatungstate is an inorganic polytungstate reagent used in analytical chemistry, density separation, and various research applications. Supplied as a solid laboratory reagent, it is commonly used to prepare density-gradient media, to separate or concentrate biological specimens, and in some biochemical assays.

  • High-density, water-soluble polytungstate reagent.
  • Used for density gradient separation and sample separation.
  • Soluble in water and DMSO with sonication (reported solubility ~50 mg/mL).
  • Available in small laboratory pack sizes such as 100 MG.
  • CAS number 12141-67-2; reported purity varies by supplier.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.