Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Metatungstate Hydrate

Catalog No. 377341250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-734-1250G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 37-734-1250G Supplier Sigma Aldrich Supplier No. 377341250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium metatungstate hydrate (NaW12O39 XH2O) can be used as a precursor for the preparation of: Tungsten oxide (WO3 x H2O) nanoplates for the electrocatalytic reaction of methanol oxidation.Tungsten carbide (WC) based counter electrodes for dye-sensitized solar cells (DSSCs).

Specifications

CAS 60583-39-3
Melting Point 180°C
Molecular Formula C46H52N4O8 -+ H2SO4 -+ 7H2O
Linear Formula (C23H26N2O4)2 -+ H2SO4 -+ 7H2O
MDL Number MFCD00150159
Quantity 25 g
Synonym Sodium polytungstate hydrate
Molecular Weight (g/mol) 1013.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.