Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium metatungstate monohydrate

Catalog No. p-4407915
Change view
Click to view available options
Quantity:
250 g
4 x 250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA2253430 250 g
AA2253456 4 x 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AA2253430 Supplier Thermo Scientific Chemicals Supplier No. 02253430
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 314075-43-9 | Na6O39W12·H2O

Sodium metatungstate monohydrate plays a vital role in sink and swim analysis. It is a derivative of sodium tungstate, which is a potential therapy for obesity. It acts as an inhibitor, which inhibits nucleoside-triphospahte- diphosphohydrolases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 314075-43-9
Molecular Formula Na6O39W12·H2O
MDL Number MFCD00061466
Synonym Sodium tungstate (meta), Sodium polytungstate

Specifications

Color White
Physical Form Powder
Quantity 250 g
Solubility Information Soluble in water.
Formula Weight 2986.13 (2968.12 Anhydrous)
Odor Odorless
Appearance White powder
Density 5.47 g/cm3
Chemical Name or Material Sodium metatungstate monohydrate
Hazard Category H302-H318-H335-H336
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P304+P340-P305+P351+P338-P310-P312-P330-P501c
EINECSNumber 412-770-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.