Learn More
Medchemexpress LLC Sodium metavanadate | 13718-26-8 | 99.3% | 1 KG
Supplier: Medchemexpress LLC HYD0852A1KG
Sodium metavanadate is a biochemical reagent used as a vanadium source in microbial cultures to study vanadium reduction in mesophilic and thermophilic methanogens. This product is for research use only.
- Biochemical reagent
- Vanadium source for microbial cultures
- Used for studying vanadium reduction in mesophilic and thermophilic methanogens