Learn More
Medchemexpress LLC Sodium metavanadate | 13718-26-8 | 99.3% | 100 G
SDP
Catalog No. 5000410435
Encompass_Preferred
Sodium metavanadate is a biochemical reagent used as a vanadium source in microbial cultures to study vanadium reduction in mesophilic and thermophilic methanogens.
  • Biochemical reagent for research use
  • Used as a vanadium source in microbial cultures
  • Studies vanadium reduction in mesophilic and thermophilic methanogens
Catalog No. 50-004-10435 Supplier Medchemexpress LLC Supplier No. HYD0852A100G
Sodium metavanadate is a biochemical reagent used as a vanadium source in microbial cultures to study vanadium reduction in mesophilic and thermophilic methanogens.
  • Biochemical reagent for research use
  • Used as a vanadium source in microbial cultures
  • Studies vanadium reduction in mesophilic and thermophilic methanogens