Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Metavanadate

Catalog No. 590088100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
59-008-8100G 100 g
59-008-825G 25 g
59-008-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 59-008-8100G Supplier Sigma Aldrich Supplier No. 590088100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium metavanadate has been used as vanadium source in the microbial culture media to study vanadium reduction by mesophilic and thermophilic methanogens.It can be used as a precursor in the preparation of vanadium pentoxide (V2O5) coated carbon nanotubes.It is also used to synthesize corrosion-inhibitive additives for organic coatings.

Specifications

CAS 13718-26-8
Quantity 100 g
Molecular Formula NaO3V
Linear Formula NaVO3
MDL Number MFCD00011125
Synonym Sodium (meta)vanadate; Sodium trioxovanadate; Sodium vanadate(V); Sodium vanadium oxide; Sodium vanadium trioxide
Molecular Weight (g/mol) 121.93 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.