Sodium metavanadate, typically 96%, V 38% min, Alfa Aesar™

Manufacturer: Alfa Aesar 11092A1

Quantity 1kg

 View more versions of this product

Catalog No. AA11092A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13718-26-8
    Physical Form Chunks and Powder
    Assay Percent Range Typically 96%
    Molecular Formula NaO3V
    Formula Weight 121.93
    MDL Number MFCD00011125
    Synonym Sodium vanadate (meta)
    Chemical Name or Material Sodium metavanadate, V 38% min.

    Specifications

    Physical Form Chunks and Powder
    Formula Weight 121.93
    Quantity 1kg
    Color Off-white
    DOT Information DOT Class: 6.1, Packaging Group: III
    EINECS Number 237-272-7
    Molecular Formula NaO3V
    Odor Odorless
    Assay Percent Range Typically 96%
    Appearance Off-white to light orange powder
    MDL Number MFCD00011125
    CAS 13718-26-8
    Synonym Sodium vanadate (meta)
    Health Hazard 1 H301-H332-H315-H319-H335: Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Sodium metavanadate, V 38% min.

    H301-H332-H315-H319-H335: Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit