Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Methacrylate, MP Biomedicals

Catalog No. ICN218055 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218055 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218055 Supplier MP Biomedicals, Inc Supplier No. 0521805550
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5536-61-8
Molecular Formula C4H5NaO2
Molecular Weight (g/mol) 108.07
MDL Number MFCD00045886
InChI Key SONHXMAHPHADTF-UHFFFAOYSA-M
Synonym sodium methacrylate, sodium polymethacrylate, acrynax, kometa, comet, komet, osakryl s, darvan 7, daxad 30, pma-na
PubChem CID 3255932
IUPAC Name sodium 2-methylprop-2-enoate
SMILES [Na+].CC(=C)C([O-])=O

Specifications

Quantity 100 g
Chemical Name or Material Sodium Methacrylate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.