Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. S0987100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0987100G 100 g
S098725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0987100G Supplier TCI America Supplier No. S0987100G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5536-61-8
Molecular Formula C4H5NaO2
Molecular Weight (g/mol) 108.07
MDL Number MFCD00045886
InChI Key SONHXMAHPHADTF-UHFFFAOYSA-M
Synonym sodium methacrylate, sodium polymethacrylate, acrynax, kometa, comet, komet, osakryl s, darvan 7, daxad 30, pma-na
PubChem CID 3255932
IUPAC Name sodium 2-methylprop-2-enoate
SMILES [Na+].CC(=C)C([O-])=O

Specifications

Melting Point 300°C
Color White
Quantity 100 g
Formula Weight 108.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Methacrylate (stabilized with MEHQ)
EINECSNumber (2)-1027
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.