Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Methanesulfonate

Catalog No. 304506100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-450-6100G 100 g
30-450-6500G 500 g
30-450-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 30-450-6100G Supplier Sigma Aldrich Supplier No. 304506100G
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium methanesulfonate is generally used as an anion source [CH3SO3]- for synthesizing ionic liquids (PILs).

Specifications

CAS 2386-57-4
Molecular Formula CH3NaO3S
Linear Formula CH3SO3Na
MDL Number MFCD00064389
Quantity 100 g
Synonym Methanesulfonic acid sodium salt
Molecular Weight (g/mol) 118.09 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.