Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium methanethiolate

Catalog No. 7174225G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-742-25G 25 g
71-742-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-742-25G Supplier Sigma Aldrich Supplier No. 7174225G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Strong nucleophile used for the synthesis of methyl aryl sulfides from halo-arenes; Alkanethiolates are efficient reagents for SN2 dealkylation of esters and aryl ethers

Specifications

Molecular Formula CH3NaS
Linear Formula CH3SNa
MDL Number MFCD00174316
Quantity 25 g
Synonym Methanethiol sodium salt; Sodium thiomethoxide
Molecular Weight (g/mol) 70.09
Percent Purity ≥90% (RT)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.