Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Methoxide, 0.22 Normal, Unstandardized (Sodium Methylate, 0.22 Molar), Reagents

Catalog No. 01333823
Change view
Click to view available options
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-823 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-823 Supplier Reagents Holdings Llc Supplier No. CS1058004C
Add to Cart
Add to Cart

Sodium Methoxide, 0.22 Normal, Unstandardized (Sodium Methylate, 0.22 Molar), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Methoxide, 0.22 Normal, Unstandardized (Sodium Methylate, 0.22 Molar)
Boiling Point 65°C
Color Colorless to Yellow
CAS 67-56-1, 124-41-4
Physical Form Liquid
Quantity 4 L
Synonym Sodium Methylate, Sodium Methanolate, Methoxysodium
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.