Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Methoxide

Catalog No. 164992500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-499-2500G 500 g
16-499-25G 5 g
16-499-2100G 100 g
16-499-22KG 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 16-499-2500G Supplier Sigma Aldrich Supplier No. 164992500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium methoxide is a sodium alkoxide. Its reaction with unactivated aryl bromides in the presence of copper catalyst under various reaction conditions has been studied. It reacts with aryl halides in the presence of a palladium catalyst in DMF to afford arenes.Sodium methoxide may be used as an alkali catalyst for the production of fatty acid methyl esters (biodiesel) from vegetable oils via transesterification.

Specifications

CAS 124-41-4
Molecular Formula CH3NaO
Linear Formula NaOCH3
MDL Number MFCD00012179
Quantity 500 g
Synonym Sodium methylate
Molecular Weight (g/mol) 54.02 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.