Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium methoxide, ca 30% w/w in methanol

Catalog No. p-7026078
Change view
Click to view available options
Quantity:
500 mL
2500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB21872AP 500 mL
AAB218720F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB21872AP Supplier Thermo Scientific Chemicals Supplier No. B21872AP
Only null left
Add to Cart

CAS: 124-41-4 | CH3NaO | 54.024 g/mol

Sodium methoxide, ca 30% w/w in methanol is used in the preparation of high-spin hexanuclear Mn(III) cluster, [NaMn6 (OMe)12 (dbm)6]+. It is useful in the synthesis of pharmaceuticals and agrochemicals. It serves as an initiator of anionic addition polymerization with ethylene oxide, forming a polyether with high molecular weight.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium methoxide, ca 30% w/w in methanol is used in the preparation of high-spin hexanuclear Mn(III) cluster, [NaMn6 (OMe)12 (dbm)6]+. It is useful in the synthesis of pharmaceuticals and agrochemicals. It serves as an initiator of anionic addition polymerization with ethylene oxide, forming a polyether with high molecular weight.

Solubility
Miscible with ethanol, fats and esters. Immiscible with hydrocarbons and organic solvents.

Notes
Air, moisture and heat sensitive. Incompatible with acids, acid chlorides, acid anhydrides, oxidizing agents, alkali metals, chlorinated solvents, water and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-41-4
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.024
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Melting Point <6°C
Density 0.969
Boiling Point 93°C
Flash Point 16°C (61°F)
Odor Alcohol-like
Linear Formula NaOCH3
Quantity 500 mL
UN Number UN1289
Beilstein 3592982
Sensitivity Air and moisture sensitive
Merck Index 14,8643
Solubility Information Miscible with ethanol,fats and esters. Immiscible with hydrocarbons and organic solvents.
Formula Weight 54.02
Concentration or Composition (by Analyte or Components) ≈30% w/w in methanol
Chemical Name or Material Sodium methoxide
Show More Show Less
Hazard Category H226-H290-H301+H311+H331-H314-H335-H336-H370-H372
Hazard Statement GHS H Statement
H225-H301-H311-H331-H370-H314
Highly flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P233-P234-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: SODIUM METHYLATE SOLUTIONS
EINECSNumber 204-699-5
RTECSNumber PC3570000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.