Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Methoxide (Powder/Laboratory), Fisher Chemical

Catalog No. S335100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S335-100 100 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S335-100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 124-41-4
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.024
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Melting Point 126°C
Color Yellow
pH 13
Packaging Glass Bottle
Quantity 100 g
UN Number 1431
Formula Weight 54.01
Grade Laboratory
Physical Form Powder or Solid
Chemical Name or Material Sodium Methoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.