Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Methoxide Solution

Catalog No. 403067100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
250 mL
800 mL
4 x 100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-306-7100ML 100 mL
40-306-7250ML 250 mL
40-306-7 4 x 100 mL
40-306-7800ML 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 40-306-7100ML Supplier Sigma Aldrich Supplier No. 403067100ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium methoxide solution is composed of 0.5M sodium methoxide in 0.5N methanol. It is a widely used organic base in organic synthesis.Sodium methoxide solution may be used in the following processes:As a solvent in the synthesis of disulfides from corresponding thiols.To extract fatty acid methyl esters by transesterification process.Synthesis of (1R,2R,3R,7aS)-1,2-dibenzyloxy-3-tert-butyldiphenylsilyloxymethylpyrrolizidin-5-one.

Specifications

CAS 124-41-4
Density 0.809 g/mL at 25°C
Boiling Point 64.6°C
Molecular Formula CH3NaO
Linear Formula CH3ONa
MDL Number MFCD00012179
Quantity 100 mL
Synonym Sodium methylate
Molecular Weight (g/mol) 54.02 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.