Learn More
Specifications
Specifications
| Chemical Name or Material | Sodium Methoxide |
| CAS | 124-41-4 |
| Physical Form | Liquid |
| Quantity | 5 mL |
| Molecular Formula | CH3NaO |
| Linear Formula | CH3ONa |
| MDL Number | MFCD00012179 |
| UN Number | UN1289 - Class 3 - PG 2 - Sodium Methylate Solutions |
| Flash Point | 9.7°C (Closed Cup) |
| Synonym | Methanolic Base; Sodium Methylate |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.