Learn More
Medchemexpress LLC Sodium methylparaben | 5026-62-0 | MFCD00016470 | 99.3% | 174.13 | C8H7NaO3 | 100 G
SDP
Catalog No. 5000301012
Encompass_Preferred
Sodium methylparaben, 99% (sodium methyl 4-hydroxybenzoate) is the sodium salt of methylparaben supplied for research and analytical use as an antifungal preservative and analytical standard. It is commonly used in formulation testing and microbiological assays and shows in vitro antifungal activity against certain molds. CAS: 5026-62-0; molecular weight: 174.13.
Catalog No. 50-003-01012 Supplier Medchemexpress LLC Supplier No. HYW016806100G
Sodium methylparaben, 99% (sodium methyl 4-hydroxybenzoate) is the sodium salt of methylparaben supplied for research and analytical use as an antifungal preservative and analytical standard. It is commonly used in formulation testing and microbiological assays and shows in vitro antifungal activity against certain molds. CAS: 5026-62-0; molecular weight: 174.13.