Learn More
Strem, An Ascensus Company CAS# 10102-40-6. 100g. Sodium molybdate dihydrate, 99%. MFCD00149170
SDP
Catalog No. 5090115336
Encompass
CAS# 10102-40-6. 100g. Sodium molybdate dihydrate, 99%. MFCD00149170. Molecular Weight: 205.92 (241.95). Molecular Formula: Na2MoO4∙2H2O. Color/form: white pwdr. Strem# 93-4234. http://www.strem.com/catalog/v/93-4234/
Catalog No. 50-901-15336 Supplier Strem, An Ascensus Company Supplier No. 934234100G
CAS# 10102-40-6. 100g. Sodium molybdate dihydrate, 99%. MFCD00149170. Molecular Weight: 205.92 (241.95). Molecular Formula: Na2MoO4∙2H2O. Color/form: white pwdr. Strem# 93-4234. http://www.strem.com/catalog/v/93-4234/