Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Molybdate, Dihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Catalog No. 02004177 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-177 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-177 Supplier Avantor Supplier No. 376401
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 10102-40-6
Quantity 500 g
Molecular Formula H4MoNa2O6
MDL Number MFCD00149170
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
Molecular Weight (g/mol) 241.96
PubChem CID 16211258
ChEBI CHEBI:75213
Formula Weight 241.95
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 3.28kg
Chemical Name or Material Sodium Molybdate, Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.