Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium molybdate dihydrate500G
Catalog No. 502898530
Encompass
Sodium molybdate dihydrate (Na2MoO4.2H2O) is basic in nature. Its crystals belong to the orthorhombic crystal system and space group Pbca.
Catalog No. 50-289-8530 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 331058500G
Sodium molybdate dihydrate (Na2MoO4.2H2O) is basic in nature. Its crystals belong to the orthorhombic crystal system and space group Pbca.