Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium molybdate dihydrate5G
Catalog No. 502898483
Encompass
Sodium molybdate dihydrate (Na2MoO4.2H2O) is basic in nature. Its crystals belong to the orthorhombic crystal system and space group Pbca.
Catalog No. 50-289-8483 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3310585G
Sodium molybdate dihydrate (Na2MoO4.2H2O) is basic in nature. Its crystals belong to the orthorhombic crystal system and space group Pbca.