Learn More
Apexbio Technology LLC Sodium molybdate 7631-95-0 10g
SDP
Catalog No. 5000063661
Encompass_Preferred
Sodium molybdate is an inorganic molybdenum salt functioning as an inhibitor of acid phosphatases as well as a modulator of glucocorticoid receptor dynamics Its primary mode of action involves inhibition of various acid phosphatase enzymes including those secreted by osteoclasts thereby interfering with bone resorption processes in vitro and in vivo This molecular property allows sodium molybdate to be employed as an experimental tool in researching metabolic bone turnover and related osteoclastic activities Additionally sodium molybdate participates in the stabilization of glucocorticoid receptors by preserving the receptor in its nonactivated complex form facilitating receptor characterization and steroid hormone signaling research
Catalog No. 50-000-63661 Supplier Apexbio Technology LLC Supplier No. B784310000
Sodium molybdate is an inorganic molybdenum salt functioning as an inhibitor of acid phosphatases as well as a modulator of glucocorticoid receptor dynamics Its primary mode of action involves inhibition of various acid phosphatase enzymes including those secreted by osteoclasts thereby interfering with bone resorption processes in vitro and in vivo This molecular property allows sodium molybdate to be employed as an experimental tool in researching metabolic bone turnover and related osteoclastic activities Additionally sodium molybdate participates in the stabilization of glucocorticoid receptors by preserving the receptor in its nonactivated complex form facilitating receptor characterization and steroid hormone signaling research