Learn More
Selleck Chemical LLC Sodium Monensin 50mg 22373-78-0
SDP
Supplier:  Selleck Chemical LLC S232450MG
Encompass_Preferred
Sodium Monensin, isolated from Streptomyces cinnamonensis, is a well-known representative of naturally polyether ionophore antibiotics. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2282