Learn More
Selleck Chemical LLC Sodium Monofluorophosphate S4013-50mg
SDP
Catalog No. 503108690
Encompass_Preferred
Sodium Monofluorophosphate (NSC248) is a competitive inhibitor of pyruvate kinase and alkaline phosphatase with Ki of 3 4 mM and 69 M respectively which also irreversibly inhibits phosphorylase phosphatase with Ki of 0 5 mM
Catalog No. 50-310-8690 Supplier Selleck Chemical LLC Supplier No. S401350MG
Add to Cart
Add to Cart
Sodium Monofluorophosphate (NSC248) is a competitive inhibitor of pyruvate kinase and alkaline phosphatase with Ki of 3 4 mM and 69 M respectively which also irreversibly inhibits phosphorylase phosphatase with Ki of 0 5 mM