Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium myristate | 822-12-8 | MFCD00042653 | 98.0% | 250.35 g/mol | C14H27NaO2 | 500 G
SDP

Catalog No. 5000282646
Encompass_Preferred

Sodium myristate is the sodium salt of myristic acid, provided as a high-purity biochemical reagent and surfactant for research and industrial applications. It is commonly used as an emulsifier, synthesis intermediate, and reagent in biochemical assays. The material is supplied as a stable powder suitable for laboratory handling and formulation work.

  • High purity: 98.0% (NMR).
  • Molecular formula: C14H27NaO2.
  • Molar mass: 250.35 g/mol.
  • CAS number: 822-12-8.
  • Physical form: white to off-white powder.
  • Typical applications: surfactant, emulsifier, reagent in synthesis and biochemical assays.
  • Pack size: 500 g.

Catalog No. 50-002-82646 Supplier Medchemexpress LLC Supplier No. HYW127604500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Sodium myristate is the sodium salt of myristic acid, provided as a high-purity biochemical reagent and surfactant for research and industrial applications. It is commonly used as an emulsifier, synthesis intermediate, and reagent in biochemical assays. The material is supplied as a stable powder suitable for laboratory handling and formulation work.

  • High purity: 98.0% (NMR).
  • Molecular formula: C14H27NaO2.
  • Molar mass: 250.35 g/mol.
  • CAS number: 822-12-8.
  • Physical form: white to off-white powder.
  • Typical applications: surfactant, emulsifier, reagent in synthesis and biochemical assays.
  • Pack size: 500 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.