Alfa Aesar™ Sodium n-dodecyl sulfate, 99% (dry wt.), water <1.5%

Manufacturer: Alfa Aesar™ A1118314

Quantity 25g

 View more versions of this product

Catalog No. AAA1118314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 151-21-3
    Molecular Formula CH3(CH2)11OSO3Na
    Formula Weight 288.38
    MDL Number MFCD00036175
    UN Number UN2926
    Beilstein 3599286
    Merck Index 14,8636
    Synonym n-Dodecyl sulfate sodium salt; Sodium lauryl sulfate
    Chemical Name or Material Sodium n-dodecyl sulfate, Water <1.5%

    Specifications

    Formula Weight 288.38
    Assay Percent Range 99% (dry wt.)
    Health Hazard 3 GHS P Statement
    P210-P261-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point ≈206°C
    TSCA TSCA
    Molecular Formula CH3(CH2)11OSO3Na
    Merck Index 14,8636
    Health Hazard 1 Danger
    RTECS Number WT1050000
    MDL Number MFCD00036175
    UN Number UN2926
    Beilstein 3599286
    Health Hazard 2 GHS H Statement
    H228-H311-H302-H315-H319-H335
    Flammable solid.
    Toxic in contact with skin.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 25g
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLIDS, TOXIC, ORGANIC, N.O.S.
    CAS 151-21-3
    Synonym n-Dodecyl sulfate sodium salt; Sodium lauryl sulfate
    Chemical Name or Material Sodium n-dodecyl sulfate, Water <1.5%
    Assay 99%, (dry wt.)

    Description

    Applications
    Denatures and solubilizes proteins for electrophoresis.Sodium n-dodecyl sulfate is an anionic detergent used to function as a protein denaturant. It is mainly used in detergents for laundry with many cleaning applications. It finds application in the determination of protein molecular weight by polyacrylamide gel electrophoresis and in the analysis of hemoglobin. In food products, it is used as an emulsifying agent and a whipping aid. It is widely found in toothpastes, shampoos, shaving creams and bubble bath formulations due to its thickening effect and ability to create a lather.

    Solubility
    Slightly soluble in ethanol.

    Notes
    Incompatible with oxidizing agents.

    GHS H Statement
    H228-H311-H302-H315-H319-H335
    Flammable solid.
    Toxic in contact with skin.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P305+P351+P338-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit