Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium n-nonyl sulfate, 99%

Catalog No. AAL0641806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0641806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0641806 Supplier Thermo Scientific Chemicals Supplier No. L0641806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1072-15-7 | C9H19NaO4S | 246.297 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072-15-7
Molecular Formula C9H19NaO4S
Molecular Weight (g/mol) 246.297
MDL Number MFCD00024893
InChI Key FTWCSAMTIKSPAT-UHFFFAOYSA-M
Synonym sodium nonyl sulfate, sodium nonyl sulphate, sodium n-nonyl sulfate, sulfuric acid, monononyl ester, sodium salt, unii-ft7kj97ok6, ft7kj97ok6, sulfuric acid, monononyl ester, sodium salt 1:1, sodium n-nonyl sulphate, dsstox_cid_27487, dsstox_rid_82377
PubChem CID 23677967
IUPAC Name sodium;nonyl sulfate
SMILES CCCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Quantity 5 g
Beilstein 3748751
Formula Weight 246.3
Percent Purity 99%
Chemical Name or Material Sodium n-nonyl sulfate
Hazard Category H302+H332-H315-H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
EINECSNumber 214-002-6
RTECSNumber WT1165000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.