Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium naphthalen-1-yl phosphate hydrate | 207569-06-0 | MFCD00150614 | >95.0% | 286.13 g/mol | C10H9Na2O5P | 250 MG
SDP

Catalog No. 5000209458
Encompass_Preferred

Sodium naphthalen-1-yl phosphate hydrate is a hydrated disodium salt of naphthyl phosphate supplied as a white to off-white solid intended for use as a biochemical reagent in life science research. It is commonly used as a substrate or reagent in enzymatic assays and organic synthesis workflows.

  • appearance: white to off-white solid.
  • cas number: 207569-06-0.
  • molecular formula: C10H9Na2O5P (hydrated disodium salt).
  • available pack sizes: 100 MG, 250 MG, 500 MG, 1 G.
  • typical use: reagent or substrate in biochemical assays and research applications.

Catalog No. 50-002-09458 Supplier Medchemexpress LLC Supplier No. 250MGHYW087942
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium naphthalen-1-yl phosphate hydrate is a hydrated disodium salt of naphthyl phosphate supplied as a white to off-white solid intended for use as a biochemical reagent in life science research. It is commonly used as a substrate or reagent in enzymatic assays and organic synthesis workflows.

  • appearance: white to off-white solid.
  • cas number: 207569-06-0.
  • molecular formula: C10H9Na2O5P (hydrated disodium salt).
  • available pack sizes: 100 MG, 250 MG, 500 MG, 1 G.
  • typical use: reagent or substrate in biochemical assays and research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.