Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium naphthalen-1-yl phosphate hydrate | 207569-06-0 | MFCD00150614 | >95.0% | 286.13 g/mol | C10H9O4P·xH2O·2Na | 1 G
SDP

Catalog No. 5000208278
Encompass_Preferred

Sodium naphthalen-1-yl phosphate hydrate is a biochemical reagent used in life-science research. It is supplied as a solid, white to off-white powder and is intended for research use only.

  • Solid, white to off-white powder.
  • Used as a biochemical reagent for life-science research.
  • Available in 100 mg to 1 g pack sizes.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • CAS number: 207569-06-0; molecular weight approximately 286.13 g/mol.

Catalog No. 50-002-08278 Supplier Medchemexpress LLC Supplier No. HYW0879421G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium naphthalen-1-yl phosphate hydrate is a biochemical reagent used in life-science research. It is supplied as a solid, white to off-white powder and is intended for research use only.

  • Solid, white to off-white powder.
  • Used as a biochemical reagent for life-science research.
  • Available in 100 mg to 1 g pack sizes.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • CAS number: 207569-06-0; molecular weight approximately 286.13 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.