Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium naphthalen-1-yl phosphate hydrate | 207569-06-0 | MFCD00150614 | C10H9O4P·xH2O·2Na | 500 MG
SDP

Catalog No. 5000208430
Encompass_Preferred

Naphthalen-1-yl phosphate sodium hydrate is a biochemical reagent intended for life science research use only. It is provided as a white to off-white solid and is supplied in multiple small pack sizes for laboratory use.

  • Intended for research use only; not for human or clinical applications.
  • Available in small pack sizes suitable for laboratory use (including 500 mg).
  • Solid appearance and white to off-white color for ease of handling and visual inspection.
  • Contains sodium counterions and hydration; chemical formula reported as C10H9O4P·xH2O·2Na.
  • Storage guidance: powder at -20°C for long-term storage or 4°C for shorter term; in solution store at -80°C or -20°C depending on duration.
  • Shipping: typically at room temperature within continental US; may vary elsewhere.

Catalog No. 50-002-08430 Supplier Medchemexpress LLC Supplier No. HYW087942500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Naphthalen-1-yl phosphate sodium hydrate is a biochemical reagent intended for life science research use only. It is provided as a white to off-white solid and is supplied in multiple small pack sizes for laboratory use.

  • Intended for research use only; not for human or clinical applications.
  • Available in small pack sizes suitable for laboratory use (including 500 mg).
  • Solid appearance and white to off-white color for ease of handling and visual inspection.
  • Contains sodium counterions and hydration; chemical formula reported as C10H9O4P·xH2O·2Na.
  • Storage guidance: powder at -20°C for long-term storage or 4°C for shorter term; in solution store at -80°C or -20°C depending on duration.
  • Shipping: typically at room temperature within continental US; may vary elsewhere.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.