Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sodium nitrate, 99+%, for biochemistry

Catalog No. AC446812500
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446812500 250 g Glass bottle
AC446810010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446812500 Supplier Thermo Scientific Chemicals Supplier No. 446812500
  • $76.00
    $63.65 / Each of 1

    Save $12.35

    16% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7631-99-4 ∣ NNaO3 ∣ 84.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
PubChem CID 24268
ChEBI CHEBI:63005
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Boiling Point 380.0°C
Melting Point 306.0°C
Physical Form Crystalline Powder and/or Crystals
Quantity 250 g
Assay Percent Range 99+%
Linear Formula NaNO3
Merck Index 15,8778
Solubility Information Solubility in water: 900g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
IUPAC Name sodium;nitrate
Formula Weight 84.99
Percent Purity ≥99%
Grade Biochemical
Packaging Glass bottle
Chemical Name or Material Sodium nitrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May intensify fire; oxidizer.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major
EINECSNumber 231-554-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.