Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrate ACS AR Granular, Macron™

Catalog No. MK7808125 Shop All Avantor Products
Change view
Click to view available options
POLYSTORMOR; 125g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-7808-125 POLYSTORMOR; 125g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-7808-125 Supplier Avantor Supplier No. 780802

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Specifications

CAS 7631-99-4
Physical Form Granules
Quantity 125 g
Molecular Formula NNaO3
MDL Number MFCD00011119
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES [Na+].[O-][N+]([O-])=O
IUPAC Name sodium nitrate
Molecular Weight (g/mol) 84.99
PubChem CID 24268
ChEBI CHEBI:63005
Formula Weight 84.99
Grade ACS
Packaging POLYSTORMOR
Chemical Name or Material Sodium Nitrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.