Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrate, ACS Reagent, Reagents

Catalog No. 01337712
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-712 100 g
01-337-711 10 kg
01-337-710 2.5 kg
01-337-709 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-712 Supplier Reagents Holdings Llc Supplier No. C1320500100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Nitrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym Nitrate of Soda
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Boiling Point 380°C
Melting Point 306°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Percent Purity 99%
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.26
Chemical Name or Material Sodium Nitrate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.