Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrate, Crystal, Reagent, ACS, 99%, Spectrum™ Chemical
SDP

Catalog No. 18606665 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-665 50 kg Poly Bottle
18-606-658 25 g Poly Bottle
18-606-659 125 g Poly Bottle
18-606-660 500 g Poly Bottle
18-606-661 6 x 500 g Poly Bottle
18-606-663 4 x 2.5 kg Poly Bottle
18-606-664 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-606-665 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S134550KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1345, 7631-99-4, NaNO3

Spectrum™ Chemical Sodium Nitrate, Crystal, Reagent, ACS , is a white solid that is particularly soluble in water and in used in the production of fertilizers, gunpowder, glass, pottery enamels, solid rocket propellant and others. It is also notably used in the production of food as a food additive. As an ACS grade Reagent, Spectrum Chemical manufactured sodium nitrate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O

Specifications

pH 5.5 to 8.3
CAS Max % 1
Physical Form Crystals or Needles
Quantity 50 kg
Percent Purity 99%
Grade ACS/Reagent
Packaging Poly Bottle
Chemical Name or Material Sodium Nitrate, Crystal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.