Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrate, Crystals, Extra Pure, MilliporeSigma™

Catalog No. M1065359050 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Fiber Carton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1065359050 50 kg Fiber Carton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1065359050 Supplier MilliporeSigma Supplier No. 1065359050
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7631-99-4
Melting Point 308°C

Specifications

Color Colorless
pH 5.5 to 8.0 (50g/L, H2O, 20°C)
Physical Form Solid
Quantity 50 kg
Molecular Formula NNaO3
MDL Number MFCD00011119
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
Solubility Information 874g/L
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
SMILES [Na+].[O-][N+]([O-])=O
IUPAC Name sodium nitrate
Molecular Weight (g/mol) 84.99
PubChem CID 24268
ChEBI CHEBI:63005
Percent Purity 99.0 to 100.5%
Packaging Fiber Carton
Density 2.26 g/3 (20 °C)
Show More Show Less
EINECSNumber 231-554-3
RTECSNumber WC5600000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.